Assay Details
| Gene Symbol: Caly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838686.3 | XP_004838743.1 | 2 - 3 | 303 | 64 | |
| XM_004838687.3 | XP_004838744.1 | 2 - 3 | 304 | 64 | ||
More Information
Set Membership: |
|

Caly
Genomic Map