Assay Details
| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836428.3 | XP_004836485.1 | 4 - 5 | 1064 | 81 | |
| XM_013078549.2 | XP_012934003.1 | 4 - 5 | 1064 | 81 | ||
| XM_013078554.2 | XP_012934008.1 | 5 - 6 | 586 | 81 | ||
More Information
Set Membership: |
|

Gabrg2
Genomic Map