Assay Details
| Gene Symbol: Apool | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868882.3 | XP_004868939.1 | 7 - 8 | 770 | 79 | |
| XM_013067111.2 | XP_012922565.1 | 7 - 8 | 624 | 79 | ||
| XM_013067112.2 | XP_012922566.2 | 7 - 8 | 660 | 79 | ||
| XM_021247657.1 | XP_021103316.1 | 8 - 9 | 781 | 79 | ||
| XM_021247658.1 | XP_021103317.1 | 8 - 9 | 742 | 79 | ||
More Information
Set Membership: |
|

Apool
Genomic Map