Assay Details
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869399.3 | XP_004869456.1 | 13 - 14 | 1680 | 60 | |
| XM_004869400.3 | XP_004869457.1 | 14 - 15 | 1617 | 60 | ||
More Information
Set Membership: |
|

Nop2
Genomic Map