Assay Details
| Gene Symbol: Nfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865803.3 | XP_004865860.1 | 3 - 4 | 789 | 63 | |
| XM_004865806.3 | XP_004865863.1 | 3 - 4 | 789 | 63 | ||
| XM_004865807.3 | XP_004865864.1 | 3 - 4 | 789 | 63 | ||
| XM_013065883.2 | XP_012921337.1 | 3 - 4 | 670 | 63 | ||
| XM_021245165.1 | XP_021100824.1 | 3 - 4 | 1175 | 63 | ||
| XM_021245166.1 | XP_021100825.1 | 3 - 4 | 1175 | 63 | ||
| XM_021245167.1 | XP_021100826.1 | 3 - 4 | 1175 | 63 | ||
| XM_021245168.1 | XP_021100827.1 | 3 - 4 | 833 | 63 | ||
More Information
Set Membership: |
|

Nfic
Genomic Map