Assay Details
| Gene Symbol: Ndufs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851822.3 | XP_004851879.2 | 2 - 3 | 227 | 53 | |
| XM_021264171.1 | XP_021119830.1 | 2 - 3 | 228 | 53 | ||
More Information
Set Membership: |
|

Ndufs3
Genomic Map