Assay Details
| Gene Symbol: Dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874996.2 | XP_004875053.1 | 2 - 3 | 302 | 66 | |
| XM_021252305.1 | XP_021107964.1 | 2 - 3 | 304 | 66 | ||
More Information
Set Membership: |
|

Dnase1l1
Genomic Map