Assay Details
| Gene Symbol: Inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856283.3 | XP_004856340.1 | 1 - 2 | 203 | 90 | |
| XM_021237624.1 | XP_021093283.1 | 1 - 2 | 203 | 90 | ||
More Information
Set Membership: |
|

Inmt
Genomic Map