Assay Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840781.3 | XP_004840838.1 | 11 - 12 | 1330 | 66 | |
| XM_004840782.3 | XP_004840839.1 | 10 - 11 | 1068 | 66 | ||
| XM_021255022.1 | XP_021110681.1 | 11 - 12 | 1238 | 66 | ||
More Information
Set Membership: |
|

Cds2
Genomic Map