Assay Details
Gene Symbol: LOC101713485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872186.3 | XP_004872243.1 | 21 - 22 | 3630 | 92 | |
XM_004872187.3 | XP_004872244.1 | 21 - 22 | 3566 | 92 | ||
XM_004872188.3 | XP_004872245.1 | 21 - 22 | 3631 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
