Assay Details
| Gene Symbol: Rpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862735.3 | XP_004862792.1 | 3 - 4 | 481 | 101 | |
| XM_021242862.1 | XP_021098521.1 | 3 - 4 | 511 | 101 | ||
More Information
Set Membership: |
|

Rpa3
Genomic Map