Assay Details
| Gene Symbol: Ilf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871424.3 | XP_004871481.1 | 11 - 12 | 967 | 57 | |
| XM_021249337.1 | XP_021104996.1 | 10 - 11 | 759 | 57 | ||
More Information
Set Membership: |
|

Ilf2
Genomic Map