Assay Details
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857808.3 | XP_004857865.1 | 4 - 5 | 920 | 75 | |
| XM_004857809.3 | XP_004857866.1 | 4 - 5 | 920 | 75 | ||
| XM_013077036.2 | XP_012932490.1 | 4 - 5 | 606 | 75 | ||
More Information
Set Membership: |
|

Azi2
Genomic Map