Assay Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845752.2 | XP_004845809.1 | 14 - 15 | 2125 | 60 | |
| XM_004845753.2 | XP_004845810.1 | 13 - 14 | 1873 | 60 | ||
| XM_013072572.2 | XP_012928026.1 | 14 - 15 | 2020 | 60 | ||
| XM_021259094.1 | XP_021114753.1 | 14 - 15 | 2128 | 60 | ||
| XM_021259095.1 | XP_021114754.1 | 14 - 15 | 2065 | 60 | ||
| XM_021259096.1 | XP_021114755.1 | 14 - 15 | 2023 | 60 | ||
| XM_021259097.1 | XP_021114756.1 | 13 - 14 | 1981 | 60 | ||
| XM_021259098.1 | XP_021114757.1 | 10 - 11 | 1438 | 60 | ||
| XM_021259099.1 | XP_021114758.1 | 10 - 11 | 1435 | 60 | ||
| XM_021259100.1 | XP_021114759.1 | 8 - 9 | 1270 | 60 | ||
| XM_021259101.1 | XP_021114760.1 | 7 - 8 | 1124 | 60 | ||
More Information
Set Membership: |
|

Sox5
Genomic Map