Assay Details
| Gene Symbol: Cpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838657.3 | XP_004838714.1 | 8 - 9 | 1437 | 60 | |
| XM_004838658.3 | XP_004838715.1 | 7 - 8 | 1257 | 60 | ||
| XM_021253197.1 | XP_021108856.1 | 8 - 9 | 1458 | 60 | ||
More Information
Set Membership: |
|

Cpn1
Genomic Map