Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864786.3 | XP_004864843.1 | 3 - 4 | 761 | 68 | |
| XM_004864787.3 | XP_004864844.1 | 3 - 4 | 779 | 68 | ||
More Information
Set Membership: |
|

Nmral1
Genomic Map