Assay Details
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837962.3 | XP_004838019.1 | 3 - 4 | 380 | 90 | |
XM_004837963.3 | XP_004838020.1 | 2 - 3 | 175 | 90 | ||
XM_004837964.3 | XP_004838021.1 | 3 - 4 | 380 | 90 | ||
XM_004837965.3 | XP_004838022.1 | 3 - 4 | 380 | 90 | ||
XM_004837967.3 | XP_004838024.1 | 3 - 4 | 380 | 90 | ||
XM_021252192.1 | XP_021107851.1 | 3 - 4 | 380 | 90 | ||
XM_021252193.1 | XP_021107852.1 | 3 - 4 | 380 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
