Assay Details
| Gene Symbol: Exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849105.2 | XP_004849162.1 | 3 - 4 | 299 | 65 | |
| XM_013073903.2 | XP_012929357.1 | 3 - 4 | 341 | 65 | ||
More Information
Set Membership: |
|

Exosc2
Genomic Map