Assay Details
| Gene Symbol: Dab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848584.3 | XP_004848641.1 | 11 - 12 | 1752 | 68 | |
| XM_021261468.1 | XP_021117127.1 | 11 - 12 | 1736 | 68 | ||
| XM_021261469.1 | XP_021117128.1 | 11 - 12 | 1736 | 68 | ||
| XM_021261470.1 | XP_021117129.1 | 10 - 11 | 1673 | 68 | ||
| XM_021261471.1 | XP_021117130.1 | 11 - 12 | 1630 | 68 | ||
| XM_021261472.1 | XP_021117131.1 | 10 - 11 | 1079 | 68 | ||
More Information
Set Membership: |
|

Dab2
Genomic Map