Assay Details
| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871995.3 | XP_004872052.1 | 8 - 9 | 1468 | 74 | |
| XM_004871996.3 | XP_004872053.1 | 8 - 9 | 1407 | 74 | ||
| XM_013068207.2 | XP_012923661.1 | 7 - 8 | 1205 | 74 | ||
| XM_021249952.1 | XP_021105611.1 | 8 - 9 | 1344 | 74 | ||
More Information
Set Membership: |
|

Prickle3
Genomic Map