Assay Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836103.3 | XP_004836160.1 | 8 - 9 | 829 | 85 | |
| XM_004836104.2 | XP_004836161.1 | 8 - 9 | 828 | 85 | ||
| XM_004836106.3 | XP_004836163.1 | 8 - 9 | 641 | 85 | ||
| XM_021236902.1 | XP_021092561.1 | 9 - 10 | 950 | 85 | ||
| XM_021236904.1 | XP_021092563.1 | 9 - 10 | 914 | 85 | ||
| XM_021236908.1 | XP_021092567.1 | 9 - 10 | 949 | 85 | ||
| XM_021236910.1 | XP_021092569.1 | 4 - 5 | 3185 | 85 | ||
| XM_021236912.1 | XP_021092571.1 | 6 - 7 | 544 | 85 | ||
More Information
Set Membership: |
|

Ccnt2
Genomic Map