Assay Details
| Gene Symbol: LOC101719002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846919.3 | XP_004846976.1 | 8 - 9 | 1048 | 86 | |
| XM_013072872.2 | XP_012928326.1 | 8 - 9 | 1048 | 86 | ||
More Information
Set Membership: |
|

LOC101719002
Genomic Map