Assay Details
| Gene Symbol: Mtss1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842884.3 | XP_004842941.1 | 11 - 12 | 1099 | 61 | |
| XM_004842885.3 | XP_004842942.1 | 10 - 11 | 994 | 61 | ||
| XM_013071498.2 | XP_012926952.1 | 10 - 11 | 1090 | 61 | ||
| XM_013071499.2 | XP_012926953.1 | 11 - 12 | 1003 | 61 | ||
| XM_013071500.2 | XP_012926954.1 | 5 - 6 | 773 | 61 | ||
More Information
Set Membership: |
|

Mtss1l
Genomic Map