Assay Details
| Gene Symbol: Dhtkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861808.2 | XP_004861865.1 | 8 - 9 | 1781 | 59 | |
| XM_004861809.1 | XP_004861866.1 | 8 - 9 | 1758 | 59 | ||
| XM_021242064.1 | XP_021097723.1 | 9 - 10 | 1715 | 59 | ||
| XM_021242065.1 | XP_021097724.1 | 8 - 9 | 1758 | 59 | ||
More Information
Set Membership: |
|

Dhtkd1
Genomic Map