Assay Details
| Gene Symbol: Rbks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839224.3 | XP_004839281.1 | 6 - 7 | 702 | 64 | |
| XM_013070146.2 | XP_012925600.1 | 6 - 7 | 705 | 64 | ||
| XM_013070148.2 | XP_012925602.1 | 6 - 7 | 704 | 64 | ||
| XM_013070149.2 | XP_012925603.1 | 6 - 7 | 703 | 64 | ||
| XM_021253645.1 | XP_021109304.1 | 6 - 7 | 707 | 64 | ||
More Information
Set Membership: |
|

Rbks
Genomic Map