Assay Details
| Gene Symbol: Tirap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242771.1 | XP_021098430.1 | 5 - 6 | 885 | 90 | |
| XM_021242772.1 | XP_021098431.1 | 3 - 4 | 476 | 90 | ||
| XM_021242773.1 | XP_021098432.1 | 4 - 5 | 769 | 90 | ||
| XM_021242774.1 | XP_021098433.1 | 4 - 5 | 592 | 90 | ||
| XM_021242775.1 | XP_021098434.1 | 5 - 6 | 1012 | 90 | ||
| XM_021242776.1 | XP_021098435.1 | 5 - 6 | 1373 | 90 | ||
| XM_021242777.1 | XP_021098436.1 | 3 - 4 | 293 | 90 | ||
| XM_021242778.1 | XP_021098437.1 | 2 - 3 | 242 | 90 | ||

Tirap
Genomic Map