Assay Details
Gene Symbol: LOC101702319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067620.2 | XP_012923074.1 | 11 - 12 | 1784 | 114 | |
XM_013067621.2 | XP_012923075.1 | 11 - 12 | 1691 | 114 | ||
XM_013067623.2 | XP_012923077.1 | 4 - 5 | 633 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
