Assay Details
| Gene Symbol: LOC101702319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067620.2 | XP_012923074.1 | 28 - 29 | 3949 | 110 | |
| XM_013067621.2 | XP_012923075.1 | 28 - 29 | 3856 | 110 | ||
| XM_013067623.2 | XP_012923077.1 | 21 - 22 | 2798 | 110 | ||
More Information
Set Membership: |
|

LOC101702319
Genomic Map