Assay Details
| Gene Symbol: LOC101702319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067620.2 | XP_012923074.1 | 29 - 30 | 4046 | 104 | |
| XM_013067621.2 | XP_012923075.1 | 29 - 30 | 3953 | 104 | ||
| XM_013067623.2 | XP_012923077.1 | 22 - 23 | 2895 | 104 | ||
More Information
Set Membership: |
|

LOC101702319
Genomic Map