Assay Details
| Gene Symbol: LOC101702319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067620.2 | XP_012923074.1 | 2 - 3 | 343 | 91 | |
| XM_013067621.2 | XP_012923075.1 | 2 - 3 | 341 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013067620.2, XM_013067621.2 transcripts.
Set Membership: |
|

LOC101702319
Genomic Map