Assay Details
| Gene Symbol: LOC101702319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067620.2 | XP_012923074.1 | 6 - 7 | 911 | 79 | |
| XM_013067621.2 | XP_012923075.1 | 6 - 7 | 818 | 79 | ||
More Information
Set Membership: |
|

LOC101702319
Genomic Map