Assay Details
| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856406.2 | XP_004856463.1 | 4 - 5 | 2523 | 65 | |
| XM_004856408.2 | XP_004856465.1 | 4 - 5 | 2523 | 65 | ||
| XM_021237577.1 | XP_021093236.1 | 4 - 5 | 2523 | 65 | ||
More Information
Set Membership: |
|

Fscn3
Genomic Map