Assay Details
| Gene Symbol: LOC101700350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859768.2 | XP_004859825.1 | 1 - 2 | 441 | 61 | |
| XM_004859769.2 | XP_004859826.1 | 1 - 2 | 444 | 61 | ||
| XM_021239941.1 | XP_021095600.1 | 1 - 2 | 443 | 61 | ||
| XM_021239942.1 | XP_021095601.1 | 1 - 2 | 443 | 61 | ||
| XM_021239943.1 | XP_021095602.1 | 1 - 2 | 445 | 61 | ||
| XM_021239944.1 | XP_021095603.1 | 1 - 2 | 447 | 61 | ||
More Information
Set Membership: |
|

LOC101700350
Genomic Map