Assay Details
| Gene Symbol: Ptpdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870058.3 | XP_004870115.1 | 3 - 4 | 800 | 82 | |
| XM_013067585.2 | XP_012923039.1 | 4 - 5 | 706 | 82 | ||
| XM_021248474.1 | XP_021104133.1 | 3 - 4 | 830 | 82 | ||
| XM_021248475.1 | XP_021104134.1 | 3 - 4 | 809 | 82 | ||
| XM_021248476.1 | XP_021104135.1 | 3 - 4 | 822 | 82 | ||
| XM_021248477.1 | XP_021104136.1 | 4 - 5 | 706 | 82 | ||
| XM_021248478.1 | XP_021104137.1 | 3 - 4 | 828 | 82 | ||
| XM_021248479.1 | XP_021104138.1 | 3 - 4 | 830 | 82 | ||
| XM_021248480.1 | XP_021104139.1 | 3 - 4 | 832 | 82 | ||
More Information
Set Membership: |
|

Ptpdc1
Genomic Map