Assay Details
| Gene Symbol: CUNH20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867834.3 | XP_004867891.2 | 2 - 3 | 385 | 87 | |
| XM_004867835.3 | XP_004867892.1 | 2 - 3 | 416 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH20orf24| Gene Symbol: CUNH20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867834.3 | XP_004867891.2 | 2 - 3 | 385 | 87 | |
| XM_004867835.3 | XP_004867892.1 | 2 - 3 | 416 | 87 | ||