Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254664.1 | XP_021110323.1 | 1 - 2 | 648 | 76 | |
| XM_021254670.1 | XP_021110329.1 | 1 - 2 | 648 | 76 | ||
| XM_021254671.1 | XP_021110330.1 | 1 - 2 | 648 | 76 | ||
| XM_021254677.1 | XP_021110336.1 | 1 - 2 | 218 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021254671.1 transcripts.
Set Membership: |
|

LOC101724144
Genomic Map