Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254664.1 | XP_021110323.1 | 2 - 3 | 731 | 60 | |
| XM_021254665.1 | XP_021110324.1 | 2 - 3 | 908 | 60 | ||
| XM_021254666.1 | XP_021110325.1 | 2 - 3 | 593 | 60 | ||
| XM_021254667.1 | XP_021110326.1 | 2 - 3 | 255 | 60 | ||
| XM_021254670.1 | XP_021110329.1 | 2 - 3 | 731 | 60 | ||
| XM_021254671.1 | XP_021110330.1 | 2 - 3 | 731 | 60 | ||
| XM_021254677.1 | XP_021110336.1 | 2 - 3 | 301 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021254671.1 transcripts.
Set Membership: |
|

LOC101724144
Genomic Map