Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254663.1 | XP_021110322.1 | 5 - 6 | 1682 | 81 | |
| XM_021254664.1 | XP_021110323.1 | 3 - 4 | 890 | 81 | ||
| XM_021254665.1 | XP_021110324.1 | 3 - 4 | 1067 | 81 | ||
| XM_021254666.1 | XP_021110325.1 | 3 - 4 | 752 | 81 | ||
| XM_021254667.1 | XP_021110326.1 | 3 - 4 | 414 | 81 | ||
| XM_021254668.1 | XP_021110327.1 | 4 - 5 | 1610 | 81 | ||
| XM_021254671.1 | XP_021110330.1 | 3 - 4 | 890 | 81 | ||
| XM_021254672.1 | XP_021110331.1 | 4 - 5 | 1610 | 81 | ||
| XM_021254673.1 | XP_021110332.1 | 3 - 4 | 304 | 81 | ||
| XM_021254674.1 | XP_021110333.1 | 2 - 3 | 247 | 81 | ||
| XM_021254676.1 | XP_021110335.1 | 2 - 3 | 247 | 81 | ||
| XM_021254677.1 | XP_021110336.1 | 3 - 4 | 460 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021254676.1 transcripts.
Set Membership: |
|

LOC101724144
Genomic Map