Assay Details
| Gene Symbol: LOC101724144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254663.1 | XP_021110322.1 | 6 - 7 | 1766 | 91 | |
| XM_021254664.1 | XP_021110323.1 | 4 - 5 | 974 | 91 | ||
| XM_021254665.1 | XP_021110324.1 | 4 - 5 | 1151 | 91 | ||
| XM_021254666.1 | XP_021110325.1 | 4 - 5 | 836 | 91 | ||
| XM_021254667.1 | XP_021110326.1 | 4 - 5 | 498 | 91 | ||
| XM_021254668.1 | XP_021110327.1 | 5 - 6 | 1694 | 91 | ||
| XM_021254676.1 | XP_021110335.1 | 3 - 4 | 331 | 91 | ||
| XM_021254677.1 | XP_021110336.1 | 4 - 5 | 544 | 91 | ||
More Information
Set Membership: |
|

LOC101724144
Genomic Map