Assay Details
| Gene Symbol: Lamtor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866281.2 | XP_004866338.1 | 6 - 7 | 486 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamtor3| Gene Symbol: Lamtor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866281.2 | XP_004866338.1 | 6 - 7 | 486 | 74 | |
Set Membership: |
|