Assay Details
| Gene Symbol: Nabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861314.3 | XP_004861371.1 | 2 - 3 | 411 | 72 | |
| XM_004861315.2 | XP_004861372.1 | 2 - 3 | 244 | 72 | ||
| XM_013078275.2 | XP_012933729.1 | 2 - 3 | 151 | 72 | ||
More Information
Set Membership: |
|

Nabp2
Genomic Map