Assay Details
| Gene Symbol: Tc2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837060.3 | XP_004837117.1 | 10 - 11 | 1335 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tc2n| Gene Symbol: Tc2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837060.3 | XP_004837117.1 | 10 - 11 | 1335 | 70 | |
Set Membership: |
|