Assay Details
| Gene Symbol: Ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848762.3 | XP_004848819.1 | 2 - 3 | 276 | 57 | |
| XM_013073840.2 | XP_012929294.1 | 2 - 3 | 158 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013073840.2 transcripts.
Set Membership: |
|

Ndor1
Genomic Map