Assay Details
| Gene Symbol: Stt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874498.3 | XP_004874555.1 | 4 - 5 | 414 | 62 | |
| XM_021251893.1 | XP_021107552.1 | 3 - 4 | 308 | 62 | ||
| XM_021251895.1 | XP_021107554.1 | 3 - 4 | 308 | 62 | ||
More Information
Set Membership: |
|

Stt3a
Genomic Map