Assay Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839103.3 | XP_004839160.1 | 5 - 6 | 691 | 63 | |
| XM_021253559.1 | XP_021109218.1 | 5 - 6 | 701 | 63 | ||
| XM_021253561.1 | XP_021109220.1 | 5 - 6 | 698 | 63 | ||
More Information
Set Membership: |
|

Cenpo
Genomic Map