Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850762.3 | XP_004850819.1 | 5 - 6 | 677 | 63 | |
| XM_004850763.2 | XP_004850820.1 | 5 - 6 | 600 | 63 | ||
| XM_004850764.2 | XP_004850821.1 | 7 - 8 | 743 | 63 | ||
| XM_004850765.3 | XP_004850822.1 | 5 - 6 | 644 | 63 | ||
| XM_004850766.3 | XP_004850823.1 | 4 - 5 | 611 | 63 | ||
| XM_021263180.1 | XP_021118839.1 | 5 - 6 | 738 | 63 | ||
| XM_021263181.1 | XP_021118840.1 | 8 - 9 | 900 | 63 | ||
| XM_021263182.1 | XP_021118841.1 | 6 - 7 | 656 | 63 | ||
| XM_021263183.1 | XP_021118842.1 | 5 - 6 | 541 | 63 | ||
| XM_021263184.1 | XP_021118843.1 | 4 - 5 | 475 | 63 | ||
| XM_021263185.1 | XP_021118844.1 | 4 - 5 | 534 | 63 | ||
| XM_021263186.1 | XP_021118845.1 | 6 - 7 | 677 | 63 | ||
| XM_021263187.1 | XP_021118846.1 | 4 - 5 | 551 | 63 | ||
| XM_021263188.1 | XP_021118847.1 | 4 - 5 | 578 | 63 | ||
| XM_021263189.1 | XP_021118848.1 | 5 - 6 | 588 | 63 | ||
| XM_021263190.1 | XP_021118849.1 | 4 - 5 | 474 | 63 | ||
| XM_021263191.1 | XP_021118850.1 | 5 - 6 | 744 | 63 | ||
| XM_021263192.1 | XP_021118851.1 | 7 - 8 | 921 | 63 | ||
| XM_021263193.1 | XP_021118852.1 | 6 - 7 | 832 | 63 | ||
| XM_021263194.1 | XP_021118853.1 | 3 - 4 | 672 | 63 | ||
More Information
Set Membership: |
|

Rcc1
Genomic Map