Assay Details
| Gene Symbol: LOC101706814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842002.3 | XP_004842059.1 | 1 - 2 | 200 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101706814| Gene Symbol: LOC101706814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842002.3 | XP_004842059.1 | 1 - 2 | 200 | 98 | |