Assay Details
| Gene Symbol: LOC101706814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842002.3 | XP_004842059.1 | 2 - 3 | 371 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101706814| Gene Symbol: LOC101706814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842002.3 | XP_004842059.1 | 2 - 3 | 371 | 61 | |
Set Membership: |
|