Assay Details
| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021264307.1 | XP_021119966.1 | 9 - 10 | 976 | 71 | |
| XM_021264308.1 | XP_021119967.1 | 7 - 8 | 774 | 71 | ||
| XM_021264309.1 | XP_021119968.1 | 7 - 8 | 808 | 71 | ||
| XM_021264310.1 | XP_021119969.1 | 9 - 10 | 979 | 71 | ||
| XM_021264311.1 | XP_021119970.1 | 7 - 8 | 795 | 71 | ||
| XM_021264312.1 | XP_021119971.1 | 4 - 5 | 4463 | 71 | ||
| XM_021264313.1 | XP_021119972.1 | 9 - 10 | 976 | 71 | ||
More Information
Set Membership: |
|

Tspan18
Genomic Map