Assay Details
| Gene Symbol: LOC101701312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857743.3 | XP_004857800.1 | 2 - 3 | 555 | 108 | |
| XM_004857744.3 | XP_004857801.1 | 2 - 3 | 555 | 108 | ||
| XM_004857745.3 | XP_004857802.1 | 2 - 3 | 555 | 108 | ||
| XM_004857746.3 | XP_004857803.1 | 2 - 3 | 555 | 108 | ||
| XM_004857747.3 | XP_004857804.1 | 2 - 3 | 555 | 108 | ||
| XM_004857749.3 | XP_004857806.1 | 2 - 3 | 555 | 108 | ||
More Information
Set Membership: |
|

LOC101701312
Genomic Map